missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL20 (MIP-3 alpha) Recombinant Protein

Human CCL20 (MIP-3 alpha) Recombinant Protein

Marke:  Gibco™ PHC1234

Additional Details : Gewicht : 0.50000kg

Artikelnummer. 10652074

  • 796.00 CHF / 1 Stück
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Human CCL20 (MIP-3 alpha) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

MIP-3-alpha (Macrophage inflammatory protein-3α) is a chemokine and is also known as CCL20.

Shipping Condition: Wet Ice.

Spezifikation

Spezifikation

Cell Culture
8 kDa
20 ug
70 aa
Store in freezer (-5 to -30°C).
< 0.1 ng/μg
Wet Ice
Chemokines & Receptors
Lyophilized
Immunology, Inflammation, Signal Transduction, Neurobiology
98 %
6364
Sequential Chromatography
MIP-3α
Human
Carrier-Free
20 μg
E. coli
Recombinant, Ligand
MIP (Macrophage Inflammatory Protein) Chemokines
Gibco™
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.