Learn More
Gibco™ Human M-CSF, AOF Recombinant Protein
Human M-CSF, AOF Recombinant Protein
Marke: Gibco™ PHG6054
Additional Details : Gewicht : 0.05400kg
Beschreibung
Human M-CSF, AOF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
M-CSF induces proliferation and differentiation of hematopoietic stem cells into macrophages and growth and differentiation of monocytes.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose dependent stimulation of proliferation of mouse M-NFS-60 cells
Shipping Condition: Wet Ice.
Spezifikation
Cell Culture | |
10 ug | |
Human | |
Carrier-Free | |
proliferation of murine M-NFS-60 cells | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
36.8 kDa | |
M-CSF AOF | |
Store in freezer (-5 to -30°C). | |
1 ng/ml | |
< 1 EU/ml | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Cardiovascular Research, Stem Cell Research, Inflammation | |
98 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.