Learn More
Gibco™ IL28B Recombinant Human Protein
Recombinant Human Interleukin-28B (IL28B) is a bioactive protein intended for use in cell culture applications
Marke: Gibco™ PHC0894
Additional Details : Gewicht : 0.00400kg
Beschreibung
IL28B is involved in the immune response, with functionality similar to type I interferons. IL28B has been shown to bind a heterodimeric receptor comprised of IL-10 receptor beta and IL-28 receptor alpha.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility—Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent protection of the cytopathic effect on A549 cells
- Proven compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Dry Ice.
Spezifikation
Cell Culture | |
10 μg | |
Human | |
Store in freezer (-5 to -30°C). | |
1 ng/ml | |
< 1 EU/ml | |
Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Virology, Drug Development |
24 kDa | |
IL-28B | |
Recombinant | |
Carrier-Free | |
Protection of the cytopathic effect on encephalomyocarditis virus challenged A549 | |
10 μg | |
Mammalian | |
Recombinant, Ligand | |
Interleukins | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.