Learn More
Invitrogen™ GFP Polyclonal Antibody

Chicken Polyclonal Antibody
Marke: Invitrogen™ PA19533
Beschreibung
PA1-9533 detects Green Fluorescence Protein in N/A samples. PA1-9533 has been successfully used in Western blot, ELISA and immunocytochemistry procedures. By Western blot, PA1-9533 detects a 32 kDa protein representing Green Fluorescence Protein. PA1-9533 immunogen is a purified recombinant green fluorescence protein.
The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Spezifikation
| GFP | |
| Polyclonal | |
| Unconjugated | |
| GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP | |
| Purified recombinant Green Fluorescent fusion Protein. | |
| 500 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Chicken | |
| IgY fraction | |
| RUO | |
| Tag | |
| Antibody | |
| IgY |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.