Learn More
Gentamicin sulfate, Tocris Bioscience™
Aminoglycoside antibiotic
Marke:  Tocris Bioscience 6442/5G
Encompass
Artikelnummer. 15943091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Gentamicin sulfate is an aminoglycoside antibiotic. Irreversibly binds bacterial ribosomes and disrupts protein synthesis. Active against gram-positive and -negative bacteria. Stable over wide range of temperatures.
Spezifikation
Spezifikation
1405-41-0
5 g