Learn More
Invitrogen™ Gentamicin Monoclonal Antibody (GB73.3), Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA110725
 Lernen Sie weitere Varianten dieses
Artikelnummer. 11508992
  • 720.00 CHF / 500 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
MA1-10725 detects Gentamicin. MA1-10725 has been successfully used in ELISA applications. The MA1-10725 immunogen is Gentamicin, conjugated to KLH.
Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Gentamicin
Monoclonal
1.3 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG2a
ELISA
GB73.3
Unconjugated
Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7
Gentamicin, conjugated to KLH.
500 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen