missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13152-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Glutathione S-transferases (GSTs) function to conjugate reduced glutathione to many exogenous and endogenous hydrophobic electrophiles. Although it shares the carboxy and amino-terminal glutathione S-transferase domains, GDAP1 (ganglioside-induced differentiation-associated protein 1) is characterized as a GST-like protein because it contains an extended GST domain II and a predicted transmembrane domain, two characteristics which are unusual for GST family members. GDAP1 may function in a signal transduction pathway that is responsible for ganglioside-induced neurite differentiation and also may play a role in protecting myelin membranes from free-radical damage. Mutations in the gene encoding GDAP1 is the cause of many forms of Charcot-Marie-Tooth disease, a common inherited disorder of the peripheral nervous system that is characterized by reduced nerve conduction velocities, slow progressive distal muscle atrophy and absent deep tendon reflexes.Spezifikation
GDAP1 | |
Polyclonal | |
Unconjugated | |
GDAP1 | |
CMTRIA, GDAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14545, 54332 | |
Antibody | |
IgG |
Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88741, Q8TB36 | |
GDAP1 | |
GDAP1 Fusion Protein Ag3769 | |
20 μL | |
Primary | |
Human, Mouse | |
Liquid |