Learn More
GBF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558092
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LTSVNKFLSY ALIDPTHEGT AEGMENMADA VTHARFVGTD PASDEVVLMK ILQVLRTLLL TPVGAHLTNE SVCEIMQSCF RICFEMRLSE LLRKS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
GBF1 Polyclonal antibody specifically detects GBF1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
GBF1 | |
Polyclonal | |
Unconjugated | |
GBF1 | |
1700083E03Rik; AI035702; ARF1GEF; AW049900; BFA-resistant GEF 1; Gbf1; golgi brefeldin A resistant guanine nucleotide exchange factor 1; golgi-specific brefeldin A-resistance factor 1; golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1; KIAA0248; mKIAA0248 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8729 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92538 | |
GBF1 | |
Recombinant protein corresponding to Human GBF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only