Learn More
GARP Monoclonal Antibody (YGIC86), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 CHF - 421.00 CHF
Spezifikation
Antigen | GARP |
---|---|
Klon | YGIC86 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983574
|
Affymetrix eBioscience
63-9891-80 |
25 μg |
178.00 CHF
25 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15993574
|
Affymetrix eBioscience
63-9891-82 |
100 μg |
421.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Description
Description: The HTA125 monoclonal antibody reacts with human Toll-like receptor 4 (TLR4). So far, at least ten members of the Toll family have been identified in humans. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR4 physically associates with another molecule called MD-2, and together with CD14, this complex is responsible for LPS recognition and signaling. TLR4 is expressed by peripheral blood monocytes. HTA125 has been reported to immunoprecipitate human TLR4 (∽100 kDa) from transfected cells. Most TLR cell surface expression, especially TLR1 and TLR4, occurs at low levels on monocytes and at even lower levels on other cell types including granulocytes and immature dendritic cells (iDC). Furthermore, a relatively high degree of variability in TLR surface expression has been reported among normal donors. Applications Reported: This HTA125 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HTA125 antibody has been pre-diluted and tested by flow cytometric analysis of cells transfected with human TLR4. This may be used at 5 μL (0.5 μg) per test.
GARP Monoclonal antibody specifically detects GARP in Mouse samples. It is validated for Flow Cytometry.Specifications
GARP | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32 | |
LRRC32 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
LRRC32 |
YGIC86 | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
G3XA59 | |
434215 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |