missing translation for 'onlineSavingsMsg'
Learn More

gamma Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen gamma Actin
Konzentration 0.16 mg/mL
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16838163
Unterlagen ansehen Aktionsdetails
Proteintech
11227-1-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16828163
Unterlagen ansehen Aktionsdetails
Proteintech
11227-1-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Actins are highly conserved proteins that are involved in various types of cell motility and in maintece of the cytoskeleton. Three main groups of actin isoforms have been identified in vertebrate animals: alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton and as mediators of internal cell motility. Actin gamma 1, encoded by this gene, is a cytoplasmic actin found in all cell types. Mutations in this gene are associated with DFNA20/26, a subtype of autosomal domit non-syndromic sensorineural progressive hearing loss and also with Baraitser-Winter syndrome. Alternative splicing results in multiple transcript variants.
Spezifikation

Spezifikation

gamma Actin
Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
P63259, P63260, P63261
11465, 287876, 71
gamma Actin Fusion Protein Ag1728
Primary
-20°C
ACTG1
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ACT, ACTB, ACTG, ACTG1, Actin, cytoplasmic 2, actin, gamma 1, Gamma actin
Actg1
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications