missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11227-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Actins are highly conserved proteins that are involved in various types of cell motility and in maintece of the cytoskeleton. Three main groups of actin isoforms have been identified in vertebrate animals: alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton and as mediators of internal cell motility. Actin gamma 1, encoded by this gene, is a cytoplasmic actin found in all cell types. Mutations in this gene are associated with DFNA20/26, a subtype of autosomal domit non-syndromic sensorineural progressive hearing loss and also with Baraitser-Winter syndrome. Alternative splicing results in multiple transcript variants.Spezifikation
gamma Actin | |
Polyclonal | |
Unconjugated | |
ACTG1 | |
ACT, ACTB, ACTG, ACTG1, Actin, cytoplasmic 2, actin, gamma 1, Gamma actin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11465, 287876, 71 | |
Antibody | |
IgG |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63259, P63260, P63261 | |
Actg1 | |
gamma Actin Fusion Protein Ag1728 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |