missing translation for 'onlineSavingsMsg'
Learn More

Galectin-9 Mouse anti-Human, PE, Clone: 9M1-3, BD Pharmingen™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 565890

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15834589

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Spezifikation

Spezifikation

Galectin-9
Monoclonal
PE
galectin 9; Gal-9; Ecalectin; HUAT; LGALS9; LEG9; Tumor antigen HOM-HD-21
Human Galectin-9 (AA 208-215) Peptide
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed.
IgG1 κ
Flow Cytometry, Flow Cytometry (Intracellular Staining)
9M1-3
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Cell Biology
Human
Purified
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.