missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Mouse anti-Human, PE, Clone: 9M1-3, BD Pharmingen™
Mouse Monoclonal Antibody
248.00 CHF - 462.00 CHF
Spezifikation
| Antigen | Galectin-9 |
|---|---|
| Klon | 9M1-3 |
| Anwendungen | Flow Cytometry, Flow Cytometry (Intracellular Staining) |
| Klassifikation | Monoclonal |
| Konjugat | PE |
Beschreibung
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .Spezifikation
| Galectin-9 | |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| PE | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Galectin-9 (AA 208-215) Peptide | |
| Primary |
| 9M1-3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| galectin 9; Gal-9; Ecalectin; HUAT; LGALS9; LEG9; Tumor antigen HOM-HD-21 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. |
For Research Use Only.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts