Learn More
GADL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559111
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CHYSMKKAAS FLGIGTENVC FVETDGRGKM IPEELEKQVW QARKEGAAPF LVC Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 58%.
GADL1 belongs to the group II decarboxylase family.Spezifikation
GADL1 | |
Polyclonal | |
Unconjugated | |
GADL1 | |
1110027M19Rik; Acidic amino acid decarboxylase GADL1; ADC; Aspartate 1-decarboxylase; CSADC; Cysteine sulfinic acid decarboxylase; GADL1; glutamate decarboxylase like 1; glutamate decarboxylase-like 1; glutamate decarboxylase-like protein 1; HuADC; HuCSADC; RGD1565145 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339896 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZQY3 | |
GADL1 | |
Recombinant protein corresponding to Human GADL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only