Learn More
Melford G-418 Disulfate Solution
Beschreibung
G-418 Disulfate, Solution is a widely used antibiotic in the life science marketplace.
In the life science field, G-418 Disulfate Solution is primarily utilized for the selection and maintenance of cells that have been genetically modified to express the neomycin resistance gene. This resistance gene allows the cells to survive in the presence of G-418, while non-modified cells are unable to survive.
Researchers and scientists working in areas such as molecular biology, genetic engineering, and biotechnology rely on G-418 Disulfate Solution for various applications. It is frequently used as a selectable marker in plasmids and other vectors to ensure that only cells carrying the desired genetic material survive and proliferate.
G-418 Disulfate Solution is compatible with a wide range of mammalian, insect, and plant cell lines, making it versatile for different cell culture systems. It is typically added to the culture medium at specific concentrations to inhibit the growth of non-resistant cells while allowing the growth of genetically modified cells.
Moreover, G-418 Disulfate Solution can be used to generate stable cell lines expressing recombinant proteins or studying gene function. Its broad-spectrum activity and compatibility with different cell types make it an important tool for researchers studying molecular mechanisms, protein expression, and genetic manipulation.
Spezifikation
Spezifikation
| Chemischer Name oder Material | G-418 Disulfate, Solution |
| CAS | 108321-42-2 |
| Menge | 20 mL |
| Summenformel | C20H40N4O10-2H2SO4 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.