Learn More
3M™ Full Face Respirators
Click to view available options
:
Model: 7907S
Beschreibung
- The popular 3M 6000 series of low maintenance respirators now includes a range of lightweight full face silicone respirators. For workers who are exposed to gases, vapours, fine particles or any combination of these.
- Lightweight
- Cost effective
- Wide field of vision
- Excellent visibility
- Easy adjustment
- Comfortable fit
- Tested in accordance with EN 140
- Easy to use bayonet fitting system
Spezifikation
Spezifikation
| Material | Silicone, Thermoplastic Elastomer |
| Produkttyp | Fullmask |
| Zur Verwendung mit (Geräte) | Twin lightweight filters or a supplied air system |