missing translation for 'onlineSavingsMsg'
Learn More

SHKBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Codice prodotto. 16800574 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
20 μL
150 μL
Dimensione della confezione:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16800574 150 μL 150µL
16810574 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16800574 Supplier Proteintech Supplier No. 127031AP150UL
 Articolo soggetto a restrizioni
Lager vorrätig
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Inhibits CBL-SH3KBP1 complex mediated down-regulation of EGFR signaling by sequestration of SH3KBP1. Binds to SH3KBP1 and prevents its interaction with CBL and inhibits translocation of SH3KBP1 to EGFR containing vesicles upon EGF stimulation.

Specifications

Antigene SHKBP1
Applicazioni Immunocytochemistry, Immunofluorescence, Western Blot
Classificazione Polyclonal
Concentrazione 0.15 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SHKBP1
N. accesso geni P0C5J9, Q6P7W2, Q8TBC3
Alias gene PP203, Sb1, SH3KBP1 binding protein 1, SHKBP1
Simboli geni SHKBP1
Specie ospite Rabbit
Immunogeno SHKBP1 Fusion Protein Ag3386
Metodo di purificazione Antigen Affinity Chromatography
Quantità 150 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 192192, 292735, 92799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Forma Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.