Learn More
Vileda Professional™ X10 LONG BUCKET WITH COLOUR CODING CLIPS 25L
For use on Controlled Environment Bucket Trolley or Pre-Prepared Trolley
Marke: Vileda Professional™ 131662
Additional Details : Gewicht : 13.72000kg
Beschreibung
- For use on the CE Bucket Trolley or the CE Pre-Prepared Trolley
- Liquid measurement lines moldest inside the bucket
- Color coding clips included to allow differentiation in colors of bucket (blue, red, yellow,green)
- Polypropylene bucket with stainless steel handle
- Autoclavable
Spezifikation
Longish Bucket | |
Gray |