Learn More
3M™ Fraser Broth Supplement BP0220010
Fraser broth supplement (5% Ferric Ammonium Citrate) is required for differential enrichment of Listeria.
Marke: 3M™ 7000053832
Additional Details : CAS : 68990-09-0 Gewicht : 0.34500kg
Beschreibung
- Flexibility of options, make what you need when you need it.
- Reduced costs.
Spezifikation
Microbiology Media | |
Environmental Test | |
10/Cs. | |
10 mL |