missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20529-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the forkhead/winged-helix family of transcription factors. It is expressed in fetal and adult brain as well as in several other organs such as the lung and gut. The protein product contains a FOX DNA-binding domain and a large polyglutamine tract and is an evolutionarily conserved transcription factor, which may bind directly to approximately 300 to 400 gene promoters in the human genome to regulate the expression of a variety of genes. This gene is required for proper development of speech and language regions of the brain during embryogenesis, and may be involved in a variety of biological pathways and cascades that may ultimately influence language development. Mutations in this gene cause speech-language disorder 1, also known as autosomal domit speech and language disorder with orofacial dyspraxia. Multiple alternative transcripts encoding different isoforms have been identified in this gene.Spezifikation
FOXP2 | |
Polyclonal | |
Unconjugated | |
FOXP2 | |
CAG repeat protein 44, CAGH44, forkhead box P2, Forkhead box protein P2, FOXP2, SPCH1, TNRC10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114142, 500037, 93986 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15409, P0CF24, P58463 | |
Foxp2 | |
FOXP2 Fusion Protein Ag14391 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |