missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19122-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
FOXO6 is a transcriptional activator.Spezifikation
FOXO6 | |
Polyclonal | |
Unconjugated | |
FOXO6 | |
forkhead box protein O6, FOXO6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100132074, 313558, 329934 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A8MYZ6, Q70KY4 | |
Foxo6 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |