missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21535-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the O class of winged helix/forkhead transcription factor family. Proteins encoded by this class are regulated by factors involved in growth and differentiation indicating they play a role in these processes. A translocation involving this gene on chromosome X and the homolog of the Drosophila trithorax gene, encoding a DNA binding protein, located on chromosome 11 is associated with leukemia. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
FOXO4 | |
Polyclonal | |
Unconjugated | |
FOXO4 | |
AFX, AFX1, forkhead box O4, Forkhead box protein O4, FOXO4, MLLT7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
302415, 4303, 54601 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P98177, Q9WVH3 | |
Foxo4 | |
FOXO4 Fusion Protein Ag16169 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |