Learn More
FOXM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13147-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Product No. 13147-1-AP is rabbit polyclonal antibody raised against C-terminal of human FOXM1. This antibody can recognize three kinds of FOXM1,FOXM1-pp (∽95-110 kDa), FOXM1-B (isoform2,∽83 kDa) and FOXM1A (isoform4, ∽90 kDa)
The protein encoded by this gene is a major non-neuronal microtubule-associated protein. This protein contains a domain similar to the microtubule-binding domains of neuronal microtubule-associated protein (MAP2) and microtubule-associated protein tau (MAPT/TAU). This protein promotes microtubule assembly, and has been shown to counteract destabilization of interphase microtubule catastrophe promotion. Cyclin B was found to interact with this protein, which targets cell division cycle 2 (CDC2) kinase to microtubules. The phosphorylation of this protein affects microtubule properties and cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
FOXM1 | |
Polyclonal | |
Unconjugated | |
FOXM1 | |
FKHL16, forkhead box M1, Forkhead box protein M1, FOXM1, FOXM1B, HFH 11, HFH11, HNF 3, HNF 3/fork head homolog 11, INS 1, M phase phosphoprotein 2, MPHOSPH2, MPP 2, MPP2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14235, 2305, 58921 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08696, P97691, Q08050 | |
Foxm1 | |
FOXM1 Fusion Protein Ag3729 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |