missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ameisensäure: Triethylamin 1:1 Lösung, Honeywell™ Fluka ™
Click to view available options
Menge:
100 ml
500 mL
Verpackung:
Glasflasche
Glass Bottle
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Chemischer Name oder Material | Formic Acid: Triethylamine 1:1 Solution |
Farbe | Colorless |
Strukturformel | (CH3CH2)3NH+ HCOO- |
UN-Nummer | UN1760 |
Beilstein | 4150472 |
Konzentration | 2M:2M concentrate |
Formelmasse | 147.22g/mol |
Dichte | 1.01g/mL at 20°C |
Güte | HPLC |
Menge | 500 mL |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'