Learn More
Foamtec UltraSORB™ Wipers
Open-cell, soft foam structure for class ISO 5 (Class 100) applications
Marke:  Foamtec HT4699
Additional Details : Gewicht : 0.45000kg
Artikelnummer. 11913554
  • auf Anfrage / 50 Stück
Verfügbar ab: 10-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.
Fordern Sie hier einen Preis für diesen Artikel an.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Features unique pore structure allows for controlled solvent transfer to screen and metal masks to aid in removal of solder paste, conductive adhesives and thick film inks.
  • High density construction and sponge-like properties
  • Dry and low-moisture cleaning
  • Ideal for screen or stencil cleaning as well as medical device manufacturing operations
  • Static dissipative to minimize ESD risk
  • Pyrogen-free for wipe-down of medical devices such as catheters
  • Features very low ionic and organic signatures
  • 100% fiber free and abrasion resistant
Spezifikation
Spezifikation
UltraSORB™ Wipes
22.86 cm
22.86 cm
For class ISO 5 (Class 100) applications, for screen or stencil cleaning as well as medical device manufacturing operations
0.27 cm
9 in.
9 in.
9 x 9 in. (22.86 x 22.86 cm)
For cleaning Metal masks used to print LTCC components, capacitors, EL, OLED, and plasma display panels