missing translation for 'onlineSavingsMsg'
Learn More

Flupyrimin, TRC

Product Code. 30405074
Change view
Click to view available options
Menge:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30405074 100 mg
30400686 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405074 Supplier Toronto Research Chemicals Supplier No. TRCF168955100MG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1689566-03-7
Summenformel C13 H9 Cl F3 N3 O
Molekulargewicht (g/mol) 315.68
Synonym Acetamide, N-[1-[(6-chloro-3-pyridinyl)methyl]-2(1H)-pyridinylidene]-2,2,2-trifluoro-, [N(E)]-, [N(E)]-N-[1-[(6-Chloro-3-pyridinyl)methyl]-2(1H)-pyridinylidene]-2,2,2-trifluoroacetamide, Flupyrimin, ME 5382
IUPAC-Name (NE)-N-[1-[(6-chloropyridin-3-yl)methyl]pyridin-2-ylidene]-2,2,2-trifluoroacetamide
SMILES FC(F)(F)C(=O)\N=C\1/C=CC=CN1Cc2ccc(Cl)nc2

Specifications

Chemischer Name oder Material Flupyrimin
Menge 100 mg
InChI-Formel InChI=1S/C13H9ClF3N3O/c14-10-5-4-9(7-18-10)8-20-6-2-1-3-11(20)19-12(21)13(15,16)17/h1-7H,8H2/b19-11+
Formelmasse 315.0386
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.