missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ FluoSpheres™ Aldehyde-Sulfate Microspheres, 1.0μm, yellow-green fluorescent (505/515), 2% solids
Beschreibung
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. Different surface modifications are available to facilitate the coupling of various molecules and proteins to the surface of the bead.
FluoSpheres™ Microsphere Specifications
Label (Ex/Em): Yellow-green (505/515)
Nominal bead diameter: 1.0μm
Coupling surface: Aldehyde-Sulfate
Solids: 2%
Characteristics of the Various FluoSpheres™ Coupling Surfaces
Carboxylate-modified FluoSpheres™ beads have a high density of pendent carboxylic acids on their surface, making them suitable for covalent coupling of proteins and other amine-containing biomolecules using water-soluble carbodiimide reagents such as EDAC
Sulfate FluoSpheres™ beads are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin
Aldehyde-sulfate FluoSpheres™ beads have surface aldehyde groups added, designed to react with proteins and other amines under very mild conditions.
Amine-modified FluoSpheres™ beads can be coupled to a wide variety of amine-reactive molecules, including the succinimidyl esters and isothiocyanates of haptens and drugs or the carboxylic acids of proteins, using a water-soluble carbodiimide
Key Applications of Microspheres
Instrument calibration (flow cytometry, microscopy, HTS, HCS)
Flow testing (microfluidics, blood flow, water flow, and air flow)
Cell biology tracers (cell differentiation and cell tracing)
Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)
Choices for FluoSpheres™ Fluorescent Microspheres
Among our complete offering of fluorescent microspheres products, you’ll find beads with these variations:
Ten fluorescent colors
Ten nominal bead diameters: 0.02μm, 0.04μm, 0.1μm, 0.2μm, 0.5μm, 1.0μm, 2.0μm, 4.0μm, 10.0μm, and 15.0μm
Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum
Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.
We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
FluoSpheres™ Microsphere Specifications
Characteristics of the Various FluoSpheres™ Coupling Surfaces
Key Applications of Microspheres
Choices for FluoSpheres™ Fluorescent Microspheres
Among our complete offering of fluorescent microspheres products, you’ll find beads with these variations:
Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.
We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
Spezifikation
Spezifikation
| Konzentration | 3.6 x 1010 particles/mL |
| Inhalt und Lagerung | 2 °C bis 8 °C |
| Inhalt | Distilled water, 2mM sodium azide |
| Beschreibung | Aldehyde-Sulfate Microspheres, 10mL, 1μm, Yellow-green Fluorescent, Excitation: 505nm, Emission Maxima: 515nm, 2% Solids, Microspheres (Also Called Latex Beads or Latex Particles) are Spherical Particles in the Colloidal Size Range That are Formed From |
| Farbe | Yellow-Green Fluorescent |
| Zur Verwendung mit (Anwendung) | Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents) |
| Material | Polystyrol |
| Partikeldichte | 1.055g/cm3 |
| Menge | 10 ml |
| Oberflächenmodifikation | Aldehyde-sulfate |
| Mehr anzeigen |
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?