Learn More
Aalborg™ 3-Channel Aluminum Flowmeter P-Style
The model Px multiple tube flow meter line offers, the convenience and simplicity of 2, 3, 4, 5 and 6 tube meters, retaining most of the unique design features associated with single tube units. Aalborg™ 3-Channel Aluminum Flowmeter P-Style provides three tube meters with 65mm flow tubes.
Marke: Aalborg™ PMR3-010073
Additional Details : Gewicht : 1.00000kg
Description
- Rib-guided or fluted metering tubes facilitate stable and accurate readings
- Interchangeability of flowtubes and floats
- Manifolding at inlet or outlet (optional)
- Ease of installation and exchange of flow tubes
- Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
- Chemical compatibility
- Simple means of panel mounting
Specifications
Flowtube | |
13.8 bar | |
65 mm |
±5% | |
3-Channel | |
121°C |