Learn More
Brady™ Untergestelle
Artikelnummer. 11712005 Alle ansehen Brady Produkte
Ansicht ändern
Click to view available options
Angezeigter Text:
ATTENTION ZONE DE TRAVAIL
ENTREE INTERDITE
Sol Sliding Attantion
Packungsgröße:
1 Stück
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Angezeigter Text Packungsgröße
11712005 ENTREE INTERDITE -
11796655 Sol Sliding Attantion -
11706665 ATTENTION ZONE DE TRAVAIL 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11712005 Lieferant Brady™ Lieferanten-Nr. 241941
  • ONLINE EXCLUSIVE PRICE
    73.15 CHF
    65.72 CHF / 1 Stück
    Sparen 7.43 CHF
    10% Rabatt
Verfügbar ab: 26-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Floor Stand designed to warn for hazardous areas, putting valuable safety information directly at the point of need.
  • These floor signs are manufactured from polyester, with a clear matt anti-slip laminate to prevent falling accidents while protecting the graphics underneath.
Spezifikation
Alternate Part Number HEAVY DUTY FLOORSTAND ENTREE INTERDIT
Angezeigter Text ENTREE INTERDITE
Höhe (angloamerikanisch) 25.98 in.
Höhe (metrisch) 660 mm
Form Rectangle
Farbe Black, Red on White
Typ Floor Stand