missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ10719 Recombinant Protein
Marke: Abnova™ H00055215-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH04277 | |
Solution | |
55215 | |
FLJ10719 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FANCI | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
33.77 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MFKDVPLTAEEVEFVVEKALSMFSKMNLQEIPPLVYQLLVLSSKGSRKSVLEGIIAFFSALDKQHNEEQSGDE | |
FLJ10719/KIAA1794 | |
FANCI | |
Wheat Germ (in vitro) | |
GST |