Learn More
Proteintech FlexAble CoraLite Plus 750 Antibody Labeling Kit for Human IgG
Alle ansehen Proteintech Produkte
Click to view available options
Menge:
10 Tests
100 Tests
50 Tests
Packungsgröße:
10 Reaktionen
200 Reaktionen
50 Reaktionen
Beschreibung
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
Spezifikation
| Menge | 10 Tests |
| Format | Liquid |