Learn More
Proteintech FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG
Artikelnummer. 18080716 Alle ansehen Proteintech Produkte
Click to view available options
Menge:
10 Tests
100 Tests
50 Tests
Packungsgröße:
10 Reaktionen
200 Reaktionen
50 Reaktionen
Artikelnummer. 18080716
Marke: Proteintech KFA10610
om dit product te kopen Registreer vandaag om een webaccount aan te maken
FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier.
FlexAble CoraLite™ Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
Menge 10 Tests
Format Liquid