Learn More
FKTN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552989
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HYLSTKNGAG LSKSKGSRIG FDSTQWRAVK KFIMLTSNQN VPVFLIDPLI LELINKNFEQ VKNTSHGSTS QCKFFCVPRD FTAFALQYHL WKNEEGWFRI AENMGFQCLK IESKDPRLDG IDSLSGTEIP LHYICKLATH AIHLV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
FKTN Polyclonal antibody specifically detects FKTN in Human samples. It is validated for Immunohistochemistry.Spezifikation
FKTN | |
Polyclonal | |
Unconjugated | |
FKTN | |
CMD1X; D830030O17Rik; Fcmd; FKTN; Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; LGMD2M; LOW QUALITY PROTEIN: fukutin; MDDGA4; MDDGB4; MDDGC4; patient fukutin; Ribitol-5-phosphate transferase; RP11-235C23.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75072 | |
FKTN | |
Recombinant protein corresponding to Human FKTN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only