missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems FITC-Labeled Human BCMA / TNFRSF17 Protein, Fc Tag DMF Filed

Marke:  ACROBiosystems BCA-HF254-25UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15826037

  • 367.32 CHF / 25 Mikrogramm


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. As a result of glycosylation, the protein migrates as 35-44 kDa under reducing (R) condition, and 60-75 kDa under non-reducing (NR) condition (SDS-PAG
Spezifikation

Spezifikation

NP_001183.2
HEK293
Human
32.0 kDa
25 µg
Met 1 - Ala 54
TNFRSF17,CD269,BCM,BCMA
FITC Labeled Protein
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Fluorescent labeled protein
Powder
>92% as determined by SDS-PAGE
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254).
-20°C, avoid light
BCMA
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation