Learn More
Duran™ Filterkerzen ohne Rohr
The Filter Candle Without Tube is suited to applications where the filtration of very small quantities is required. This filtration is possible from a liquid or a carrier gas.
auf Anfrage
Beschreibung
- Without tube
- Duran™ borosilicate glass 3.3 Can be used as a filter to separate the smallest amount of solids from the filter surface
Spezifikation
1 Ea. | |
65 mm |