missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA521640
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.
Spezifikation
| Fibulin 2 | |
| Polyclonal | |
| Unconjugated | |
| FBLN2 | |
| 5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14115, 2199, 282583 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.19 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P37889, P98095 | |
| FBLN2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Fibulin 2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction