missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblasts/Epithelial cells Antibody (D7-FIB), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB600-777AF700
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Fibroblasts/Epithelial cells Monoclonal specifically detects Fibroblasts/Epithelial cells in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Spezifikation
| Fibroblasts/Epithelial cells | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB600-777 recognizes a 112kD molecule expressed on the cell surface of human fibroblasts and epithelial cells. In peripheral blood the antibody stains myeloid cells and a very small number of lymphocytes. Studies upon the antigen have shown it to be sensitive to SDS, but resistant to trypsin, tunicamycin and collagenase. In immunohistological studies the antibody has also been found to bind to epithelium, myoepthelium, smooth muscle and some leucocytes. D7-FIB has been shown to be useful as a cell membrane marker to characterize chondrocyte differentiation giving a positive reaction with dedifferentiated human chondrocytes, and negative with differentiated chondrocytes (3). This product is routinely tested in flow cytometry on the KG1 cell line. | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| D7-FIB | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| Human foreskin fibroblasts | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur