missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB110-85534DL594
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Fibroblast Activation Protein alpha/FAP Polyclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Western Blot, ELISA
Spezifikation
| Fibroblast Activation Protein alpha/FAP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| DyLight 594 | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| This affinity purified Fibroblast Activation Protein alpha/FAP Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse Fibroblast Activation Protein alpha/Fibroblast Activation Protein alpha/FAP protein. (Uniprot: P97321) | |
| 0.1 mL | |
| Cancer, Protein Phosphatase | |
| 2191 | |
| Store at 4°C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur