missing translation for 'onlineSavingsMsg'
Learn More

Fibrinogen Polyclonal Antibody, FITC, Invitrogen™

Artikelnummer. 11315813
Change view
Click to view available options
Menge:
2 mL
Packungsgröße:
2 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11315813 2 mL 2 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11315813 Lieferant Invitrogen Lieferanten-Nr. PA516599

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

PA5-16599 targets Fibrinogen in IF applications and shows reactivity with Human samples. The PA5-16599 immunogen is fibrinogen isolated from human serum.

Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Fibrinogen
Anwendungen Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Zusammensetzung TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4
Gen FGA
Gen-Zugriffsnummer P02671
Gen-Alias fibrinogen, A alpha polypeptide
Gensymbole FGA
Wirtsspezies Rabbit
Immunogen Fibrinogen isolated from human serum
Reinigungsverfahren Purified
Menge 2 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 2243
Zielspezies Human
Inhalt und Lagerung 4° C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.