missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferritin Light Chain Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA529599
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Dog (87%), Cat (86%), Pig (88%), Rabbit (86%), Rhesus Monkey (97%), Bovine (84%), Guinea pig (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.
Spezifikation
| Ferritin Light Chain | |
| Polyclonal | |
| Unconjugated | |
| FTL | |
| ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14325, 2512, 29292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P02792, P02793, P29391 | |
| FTL, Ftl1 | |
| Recombinant protein encompassing a sequence within the center region of human Ferritin Light Chain. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur