Learn More
Versilon FEP High-Purity Tubing
Alle ansehen Cole-Parmer Produkte
Click to view available options
Innendurchmesser (angloamerikanisch):
1/16 in.
1/2 in.
1/4 in.
1/8 in.
1 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
Außendurchmesser (angloamerikanisch):
1-1/8 in
1/2 in.
1/4 in.
1/8 in.
1 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
Packungsgröße:
15 Meter
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Made from chemically inert thermoplastic. Transparent and flexible—ideal for chemical transfer applications. Withstands temperatures up to 206°C (402°F). Noncorroding, nonoxidizing, and UV resistant
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ADCF |
| Innendurchmesser (angloamerikanisch) | 1/2 in. |
| Material | FEP |
| Außendurchmesser (angloamerikanisch) | 5/8 in. |
| Gesamtlänge (metrisch) | 50 ft. |
| Farbe | Transparent |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.