Learn More
Value Plastics Female Luer Thread Style Caps
Alle ansehen Cole-Parmer Produkte
Click to view available options
Material (Dichtung):
Transparentes Polycarbonat
Weißes Nylon
Packungsgröße:
25 Stück
Beschreibung
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Thread design provides a secure closure—preventing contamination
Spezifikation
Spezifikation
| Zertifizierung/Konformität | Non-DEHP;Non-Phthalate;RoHS |
| Verpackungsmenge | 25/Pk. |
| Einlass | Female Luer Thread, Cap |
| Material (Dichtung) | Weißes Nylon |