missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDFT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13128-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF regulates the transient response of several muscle genes in response to growth factors and recruits accessory myogenic factors to activate these muscle genes. SRF is required for the formation of vertebrate mesoderm leading to the origin of the cardiovascular system. Myocardin, in association with SRF in cardiac muscle cells, activates cardiac muscle promoters. Myocardin-related transcription factors A and B (MRTF-A and MRTF-B) interact with SRF and act as stimulators for its transcriptional activity. MRTF-B is crucial for skeletal myogenic differentiation.Spezifikation
FDFT1 | |
Polyclonal | |
Unconjugated | |
FDFT1 | |
DGPT, ERG9, FDFT1, FPP:FPP farnesyltransferase, SQS, Squalene synthase, SS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14137, 2222, 29580 | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P37268, P53798, Q02769 | |
FDFT1 | |
FDFT1 Fusion Protein Ag3743 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Liquid |