missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (FI-17), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
Marke: Novus Biologicals NB100-316SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
FANCD2 Monoclonal specifically detects FANCD2 in Human, Hamster (Negative), Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Spezifikation
| FANCD2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| Mouse | |
| 164.1 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Human, Hamster (Negative), Mouse (Negative) | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
| FI-17 | |
| Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Q9BXW9 | |
| FANCD2 | |
| This FANCD2 Antibody (FI-17) was developed against human FANCD2 fusion protein [Swiss-Prot: Q9BXW9] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human. This antibody does not react with mouse or hamster. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur