missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM185A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20911-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
FAM185A | |
Polyclonal | |
Unconjugated | |
FAM185A | |
FAM185A, Protein FAM185A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
222234 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N0U4 | |
FAM185A | |
FAM185A Fusion Protein Ag15041 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |