missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF2 Mouse anti-Human, Clone: 3H6D2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66629-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Spezifikation
FAF2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96CS3 | |
FAF2 | |
FAF2 Fusion Protein Ag21536 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3H6D2 | |
Unconjugated | |
FAF2 | |
ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
Mouse | |
Protein G | |
RUO | |
23197 | |
-20°C | |
Liquid |