Learn More
FABP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16860014 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16860014 150 μL 150 Mikroliter
16870014 20 μL 20 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16860014 Lieferant Proteintech Lieferanten-Nr. 123481AP150UL
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
Fatty acid-binding proteins, designated FABPs, are a family of homologous cytoplasmic proteins that are expressed in a highly tissue-specific manner and play an integral role in the balance between lipid and carbohydrate metabolism. FABPs mediate fatty acid (FA) and/or hydrophobic ligand uptake, transport and targeting within their respective tissues. The mechanisms underlying these actions can give rise to both passive diffusional uptake and protein-mediated transmembrane transport of FAs. FABPs are expressed in adipocytes (A-FABP), brain (B-FABP), epidermis (E-FABP, also designated psoriasis-associated FABP or PA-FABP), muscle and heart (H-FABP, also designated mammary-derived growth inhibitor or MDGI), intestine (I-FABP), liver (L-FABP), myelin (M-FABP) and testis (T-FABP). Epithelial fatty acid-binding protein (E-FABP) binds stearic acid and may play a role in keratinocyte differentiation. E-FABP is upregulated in rat dorsal root ganglia after sciatic nerve crush and in differentiating neurons during development.
Spezifikation
Antigen FABP5
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 0.43 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen FABP5
Gen-Zugriffsnummer P55053, Q01469, Q05816
Gen-Alias E FABP, EFABP, FABP5, Fatty acid binding protein 5, PA FABP, PAFABP
Gensymbole FABP5
Wirtsspezies Rabbit
Immunogen FABP5 Fusion Protein Ag3005
Reinigungsverfahren Antigen Affinity Chromatography
Menge 150 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 140868, 16592, 2171
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG