missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102377
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Antibody detects endogenous levels of total FAAH.
Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
Tekniske data
| FAAH | |
| Polyclonal | |
| Unconjugated | |
| FAAH | |
| amide hydrolase; anandamide amidohydrolase 1; AW412498; FA2H; Faah; FAAH1; FAAH-1; fatty acid amide hydrolase; fatty acid amide hydrolase protein; fatty-acid amide hydrolase 1; fatty-acid amide hydrolase 1-like; LOC100911581; MGC102823; MGC138146; Oleamide hydrolase; Oleamide hydrolase 1; PSAB | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14073, 2166, 29347 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00519, O08914, P97612 | |
| FAAH | |
| A synthesized peptide derived from human FAAH(Accession O00519), corresponding to amino acid residues P546-S579. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion