missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (CL2375), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-36759
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ezrin Monoclonal specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| Ezrin | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL2375 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000-1:10000 | |
| P15311 | |
| EZR | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:REEKHQKQLERQQLETEKKRRETVEREKEQMMREKEELMLRLQDYEEKTKKAERELSEQIQRALQLEEERKRAQEEAERLEADRMAALRAKEELERQAVDQIKSQEQLAAELAEYTAKIALLE | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| 7430 | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur