missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EYA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA565034
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: SQSPGQTGFLS YGTSFSTPQP GQAPYSYQMQ GSSFTTSSGI YTGNNSLTNS SGFNSSQQDY PSYPSFGQGQ YAQYYNSSPY PAHYMTSSNT SPTTPSTNAT YQLQEPPSGI TSQAVTDPTA EYSTIHSPST PIKDSDSDRL RRGSD Highest antigen sequence identity to the following orthologs - mouse 95%, rat 97%.
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
Spezifikation
| EYA1 | |
| Polyclonal | |
| Unconjugated | |
| EYA1 | |
| BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2138 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99502 | |
| EYA1 | |
| Recombinant Human EYA1. Recombinant protein control fragment (Product #RP-105800). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur